The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018384.4 | NP_060854.2 | 2 - 3 | 402 | 103 |
GenBank mRNA | AK002158.1 | - | 2 - 3 | 338 | 103 |
AK297881.1 | - | 3 - 4 | 226 | 103 | |
AL557446.3 | - | 2 - 3 | 176 | 103 | |
BC011732.2 | - | 2 - 3 | 135 | 103 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199577.1 | NP_001186506.1 | 5 - 6 | 787 | 103 |
NM_001303630.1 | NP_001290559.1 | 4 - 5 | 428 | 103 |