Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XAGE1B
Entrez Gene ID
653220
Gene Name
X antigen family member 1B
Gene Aliases
CT12.1, CT12.1A, CT12.1B, CTP9, GAGED2, XAGE-1, XAGE1, XAGE1A
Chromosome Location
Chr.X: 52495668 - 52500815 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097594.2 NP_001091063.2 3 - 4 511 101
NM_001097596.2 NP_001091065.1 3 - 4 374 101
NR_033254.1 3 - 4 622 101
XM_017029748.1 XP_016885237.1 3 - 4 643 101
XM_017029749.1 XP_016885238.1 3 - 4 511 101

Gene Symbol
XAGE1E
Entrez Gene ID
653067
Gene Name
X antigen family member 1E
Gene Aliases
CT12.1, CT12.1C, CT12.1D, CT12.1E, CTP9, GAGED2, XAGE-1, XAGE1, XAGE1C, XAGE1D
Chromosome Location
Chr.X: 52512074 - 52517221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097604.2 NP_001091073.2 3 - 4 511 101
NM_001097605.2 NP_001091074.1 3 - 4 374 101
NR_033257.1 3 - 4 622 101
XM_017029746.1 XP_016885235.1 3 - 4 643 101
XM_017029747.1 XP_016885236.1 3 - 4 511 101
GenBank mRNA AF251237.1 - 3 - 4 503 101
AJ290447.1 - 3 - 4 347 101
AJ318878.1 - 3 - 4 622 101
AJ318879.1 - 3 - 4 363 101
BC009538.1 - 3 - 4 406 101

View Full Product Details