Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEUROD4
Entrez Gene ID
58158
Gene Name
neuronal differentiation 4
Gene Aliases
ATH-3, ATH3, Atoh3, MATH-3, MATH3, bHLHa4
Chromosome Location
Chr.12: 55019945 - 55030017 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021191.2 NP_067014.2 1 - 2 366 91
GenBank mRNA AK314136.1 - 1 - 2 281 91
DA417566.1 - 1 - 2 281 91

View Full Product Details