The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256410.1 | NP_001243339.1 | 7 - 8 | 725 | 65 |
NM_001256412.1 | NP_001243341.1 | 4 - 5 | 446 | 65 | |
NM_001256415.1 | NP_001243344.1 | 5 - 6 | 566 | 65 | |
NM_021252.4 | NP_067075.1 | 6 - 7 | 638 | 65 | |
NR_046172.1 | 5 - 6 | 576 | 65 | ||
GenBank mRNA | AB209698.1 | - | 2 - 3 | 5400 | 65 |
AF087860.1 | - | 6 - 7 | 496 | 65 | |
AF136974.1 | - | 6 - 7 | 552 | 65 | |
AF137372.1 | - | 6 - 7 | 442 | 65 | |
AF498950.1 | - | 6 - 7 | 442 | 65 | |
AK001555.1 | - | 6 - 7 | 442 | 65 | |
AK223153.1 | - | 6 - 7 | 465 | 65 | |
AK295443.1 | - | 4 - 5 | 316 | 65 | |
AK299457.1 | - | 5 - 6 | 442 | 65 | |
AL136734.1 | - | 6 - 7 | 491 | 65 | |
AY574034.1 | - | 7 - 8 | 614 | 65 | |
BC015014.2 | - | 6 - 7 | 492 | 65 | |
BC029350.1 | - | 6 - 7 | 508 | 65 | |
BP210053.1 | - | 5 - 6 | 440 | 65 | |
BT009840.1 | - | 6 - 7 | 442 | 65 | |
CR533455.1 | - | 6 - 7 | 442 | 65 |