The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023112.3 | NP_075601.1 | 3 - 4 | 373 | 83 |
XM_005268055.3 | XP_005268112.1 | 4 - 5 | 423 | 83 | |
XM_011537148.2 | XP_011535450.1 | 3 - 4 | 375 | 83 | |
GenBank mRNA | AK025569.1 | - | 3 - 4 | 349 | 83 |
AY177201.1 | - | 2 - 3 | 213 | 83 | |
BC068058.1 | - | 3 - 4 | 373 | 83 | |
CR457345.1 | - | 2 - 3 | 213 | 83 |