The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135170.1 | NP_001128642.1 | 2 - 3 | 520 | 82 |
NM_001135171.1 | NP_001128643.1 | 2 - 3 | 467 | 82 | |
NM_031911.4 | NP_114117.1 | 2 - 3 | 350 | 82 | |
XM_011513772.1 | XP_011512074.1 | 3 - 4 | 390 | 82 | |
XM_011513773.1 | XP_011512075.1 | 3 - 4 | 439 | 82 | |
GenBank mRNA | AF329839.1 | - | 2 - 3 | 325 | 82 |
AK314802.1 | - | 2 - 3 | 468 | 82 | |
BC022187.2 | - | 2 - 3 | 468 | 82 | |
BC024015.2 | - | 2 - 3 | 468 | 82 | |
DB069980.1 | - | 2 - 3 | 468 | 82 | |
DB290077.1 | - | 2 - 3 | 351 | 82 |