Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCB9
Entrez Gene ID
23457
Gene Name
ATP binding cassette subfamily B member 9
Gene Aliases
EST122234, TAPL
Chromosome Location
Chr.12: 122918708 - 122975160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243014.1 NP_001229943.1 7 - 8 1690 85
NM_019625.3 NP_062571.1 7 - 8 1690 85
NM_203444.3 NP_982269.2 7 - 8 1690 85
XM_005253558.2 XP_005253615.1 7 - 8 1630 85
XM_011538095.2 XP_011536397.1 7 - 8 1913 85
XM_011538096.2 XP_011536398.1 7 - 8 1709 85
XM_011538098.2 XP_011536400.1 6 - 7 2555 85
XM_017019103.1 XP_016874592.1 8 - 9 1751 85
GenBank mRNA AB040953.2 - 7 - 8 4219 85
AB045381.2 - 6 - 7 1439 85
AB112582.1 - 7 - 8 1641 85
AB112583.1 - 7 - 8 1641 85
AB177852.1 - 7 - 8 1602 85
AF216494.1 - 7 - 8 1678 85
AK074738.1 - 4 - 5 634 85
AK294225.1 - 1 - 2 249 85
BC017348.2 - 6 - 7 1405 85
BC064384.1 - 7 - 8 1958 85

View Full Product Details