The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012228.3 | NP_036360.3 | 2 - 3 | 330 | 100 |
XM_011519426.2 | XP_011517728.1 | 2 - 3 | 264 | 100 | |
GenBank mRNA | AF122004.1 | - | 2 - 3 | 247 | 100 |
AF151889.1 | - | 2 - 3 | 278 | 100 | |
BC066305.1 | - | 2 - 3 | 244 | 100 | |
BC117471.1 | - | 2 - 3 | 379 | 100 | |
BC130380.1 | - | 2 - 3 | 379 | 100 |