The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023918.1 | NP_076407.1 | 1 - 1 | 350 | 81 |
GenBank mRNA | BC069084.1 | - | 1 - 1 | 350 | 81 |
BC069551.1 | - | 1 - 1 | 350 | 81 | |
BC096701.1 | - | 1 - 1 | 350 | 81 | |
BC096735.1 | - | 1 - 1 | 350 | 81 | |
BC098167.1 | - | 1 - 1 | 350 | 81 | |
BC098353.1 | - | 1 - 1 | 350 | 81 |