The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142443.1 | NP_001135915.1 | 5 - 6 | 656 | 122 |
NM_001142444.1 | NP_001135916.1 | 3 - 4 | 441 | 122 | |
NM_025083.3 | NP_079359.2 | 2 - 3 | 352 | 122 | |
GenBank mRNA | AF193058.1 | - | 3 - 4 | 389 | 122 |
AK024781.1 | - | 3 - 4 | 381 | 122 | |
AK056339.1 | - | 2 - 3 | 352 | 122 | |
AK225099.1 | - | 3 - 4 | 381 | 122 | |
BC011534.1 | - | 2 - 3 | 326 | 122 | |
BC021271.2 | - | 5 - 6 | 630 | 122 | |
DA168050.1 | - | 3 - 4 | 436 | 122 |