The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025184.3 | NP_079460.2 | 8 - 9 | 1369 | 125 |
XM_006724562.2 | XP_006724625.1 | 7 - 8 | 1082 | 125 | |
GenBank mRNA | AK026496.1 | - | 8 - 9 | 1222 | 126 |
AK291187.1 | - | 8 - 9 | 1510 | 125 | |
AK292967.1 | - | 8 - 9 | 125 | ||
BC031039.1 | - | 8 - 9 | 1364 | 125 | |
CR749363.1 | - | 3 - 4 | 1030 | 125 |