The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002086.4 | NP_002077.1 | 6 - 6 | 2704 | 68 |
NM_203506.2 | NP_987102.1 | 5 - 5 | 2582 | 68 | |
GenBank mRNA | AF171699.1 | - | 1 - 1 | 282 | 68 |
AF246238.1 | - | 1 - 1 | 653 | 68 | |
AF302079.1 | - | 1 - 1 | 272 | 68 | |
AK024539.1 | - | 1 - 1 | 626 | 68 | |
BC019082.2 | - | 1 - 1 | 665 | 68 | |
BC033644.1 | - | 1 - 1 | 605 | 68 | |
BC043007.1 | - | 1 - 1 | 530 | 68 |