The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182627.1 | NP_872433.1 | 3 - 4 | 961 | 63 |
GenBank mRNA | AK314105.1 | - | 3 - 4 | 970 | 63 |
BC054030.1 | - | 3 - 4 | 961 | 63 | |
EF011620.1 | - | 3 - 4 | 961 | 63 |