The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033224.4 | NP_150093.1 | 1 - 1 | 844 | 60 |
GenBank mRNA | AY039216.1 | - | 1 - 1 | 820 | 60 |
BC101735.1 | - | 1 - 1 | 826 | 60 | |
BC101737.1 | - | 1 - 1 | 826 | 60 | |
BU177276.1 | - | 1 - 1 | 244 | 60 |