The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016337.2 | NP_057421.1 | 3 - 4 | 448 | 100 |
XM_005267749.3 | XP_005267806.1 | 3 - 4 | 1067 | 100 | |
XM_011536828.2 | XP_011535130.1 | 3 - 4 | 1081 | 100 | |
XM_017021363.1 | XP_016876852.1 | 3 - 4 | 1067 | 100 | |
XM_017021364.1 | XP_016876853.1 | 3 - 4 | 954 | 100 | |
GenBank mRNA | AF052504.1 | - | 3 - 4 | 422 | 100 |
AF087843.1 | - | 3 - 4 | 427 | 100 | |
AF112209.1 | - | 3 - 4 | 541 | 100 | |
AF131766.1 | - | 1 - 2 | 195 | 100 | |
AK289720.1 | - | 4 - 5 | 458 | 100 | |
AK295919.1 | - | 3 - 4 | 656 | 100 | |
AK301379.1 | - | 3 - 4 | 440 | 100 | |
BC023997.2 | - | 3 - 4 | 597 | 100 | |
BC032358.1 | - | 3 - 4 | 419 | 100 | |
BQ083568.1 | - | 3 - 4 | 448 | 100 |