Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLC1
Entrez Gene ID
3831
Gene Name
kinesin light chain 1
Gene Aliases
KLC, KNS2, KNS2A
Chromosome Location
Chr.14: 103629188 - 103701551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK054874.1 - 1 - 1 552 94


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB251969.1 - 1 140 Chr. 14 - 103685374 - 103685828 [+] on Build GRCh38 94
GenBankmRNA AK092888.1 - 1 2460 Chr. 14 - 103683054 - 103685894 [+] on Build GRCh38 94
GenBankmRNA BM907997.1 - 421 Chr. - - [-] on Build 94
GenBankmRNA AA508740.1 - 1 112 Chr. 14 - 103685405 - 103685717 [+] on Build GRCh38 94
GenBankmRNA DB306767.1 - 1 Chr. 14 - 103685353 - 103685894 [-] on Build GRCh38 94
GenBankmRNA DB286400.1 - 1 233 Chr. 14 - 103685281 - 103685836 [+] on Build GRCh38 94


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream apoptogenic 1, mitochondrial APOPT1 APOP, APOP1, C14orf153 -92.8kb Sense Chr. - 103562957 - 103592187 on Build GRCh38
Downstream X-ray repair cross complementing 3 XRCC3 CMM6 11.4kb Anti-sense Chr. - 103697611 - 103715486 on Build GRCh38

View Full Product Details