Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATA13
Entrez Gene ID
221178
Gene Name
spermatogenesis associated 13
Gene Aliases
ARHGEF29, ASEF2
Chromosome Location
Chr.13: 23979626 - 24307074 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153023.3 NP_694568.1 1 - 2 222 87
GenBank mRNA AK055770.1 - 1 - 2 190 87

View Full Product Details