Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TICRR
- Entrez Gene ID
-
90381
- Gene Name
- TOPBP1 interacting checkpoint and replication regulator
- Gene Aliases
-
C15orf42, SLD3, Treslin
- Chromosome Location
-
Chr.15: 89575587 - 89628023 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC002881.2 |
- |
3 - 4 |
740 |
88 |
View Full Product Details