The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030630.2 | NP_085133.1 | 1 - 2 | 218 | 93 |
XM_005257226.1 | XP_005257283.1 | 1 - 2 | 216 | 93 | |
GenBank mRNA | AK074401.1 | - | 1 - 2 | 147 | 93 |
AK095565.1 | - | 1 - 2 | 176 | 93 | |
AK304081.1 | - | 1 - 2 | 133 | 93 | |
AL137556.1 | - | 1 - 2 | 165 | 93 | |
BC035372.1 | - | 1 - 2 | 134 | 93 | |
DA336027.1 | - | 1 - 2 | 218 | 93 |