Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAMDC4
Entrez Gene ID
158056
Gene Name
MAM domain containing 4
Gene Aliases
AEGP
Chromosome Location
Chr.9: 136852367 - 136860799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL137659.1 - 1 - 2 160 98

View Full Product Details