The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152284.3 | NP_689497.1 | 1 - 2 | 367 | 70 |
GenBank mRNA | AB120734.1 | - | 1 - 2 | 199 | 70 |
AK000049.1 | - | 1 - 2 | 330 | 70 | |
AK314873.1 | - | 1 - 2 | 366 | 70 | |
AY329086.1 | - | 1 - 2 | 201 | 70 | |
BC014321.1 | - | 1 - 2 | 371 | 70 | |
BX492886.1 | - | 1 - 2 | 387 | 70 |