The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002998.3 | NP_002989.2 | 1 - 2 | 676 | 103 |
GenBank mRNA | AK025488.1 | - | 1 - 2 | 517 | 103 |
AK074530.1 | - | 1 - 2 | 462 | 103 | |
AK097839.1 | - | 2 - 3 | 477 | 103 | |
AK130131.1 | - | 1 - 2 | 517 | 103 | |
BC030133.2 | - | 1 - 2 | 662 | 103 | |
BC049836.1 | - | 1 - 2 | 394 | 103 | |
J04621.1 | - | 1 - 2 | 646 | 103 |