The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020927.2 | NP_065978.1 | 3 - 4 | 701 | 116 |
GenBank mRNA | AB046796.1 | - | 3 - 4 | 671 | 116 |
AK290153.1 | - | 3 - 4 | 679 | 116 | |
AL834240.1 | - | 2 - 3 | 316 | 116 | |
BC033537.1 | - | 3 - 4 | 679 | 116 |