The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012233.2 | NP_036365.1 | 23 - 24 | 2756 | 85 |
XM_011510823.2 | XP_011509125.1 | 23 - 24 | 2756 | 85 | |
XM_011510825.2 | XP_011509127.1 | 23 - 24 | 2756 | 85 | |
GenBank mRNA | AB449877.1 | - | 23 - 24 | 2713 | 85 |
AK022494.1 | - | 23 - 24 | 2728 | 85 | |
AK056058.1 | - | 1 - 2 | 664 | 85 | |
AK309118.1 | - | 6 - 7 | 658 | 85 | |
AL096752.1 | - | 4 - 5 | 1482 | 85 | |
BC022977.1 | - | 10 - 11 | 1400 | 85 | |
BC146809.1 | - | 25 - 26 | 2993 | 85 | |
D31886.1 | - | 23 - 24 | 2715 | 85 |