Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP4F8
Entrez Gene ID
11283
Gene Name
cytochrome P450 family 4 subfamily F member 8
Gene Aliases
CPF8, CYPIVF8
Chromosome Location
Chr.19: 15615218 - 15629638 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007253.3 NP_009184.1 10 - 11 1314 71
GenBank mRNA AF133298.1 - 9 - 10 1257 71
AK096809.1 - 6 - 7 2968 71
AK300539.1 - 6 - 7 865 71
AK314685.1 - 10 - 11 1312 71
DB329701.1 - 3 - 4 71

View Full Product Details