The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320049.1 | NP_001306978.1 | 1 - 2 | 310 | 127 |
NM_006331.7 | NP_006322.4 | 1 - 2 | 310 | 127 | |
NR_135131.1 | 1 - 2 | 310 | 127 | ||
GenBank mRNA | AA314580.1 | - | 1 - 2 | 158 | 126 |
AK291597.1 | - | 1 - 2 | 175 | 127 | |
AK298207.1 | - | 1 - 2 | 178 | 127 | |
AK298745.1 | - | 1 - 2 | 170 | 127 | |
AK307177.1 | - | 1 - 2 | 175 | 127 | |
BC055314.1 | - | 1 - 2 | 202 | 127 | |
BM709304.1 | - | 1 - 2 | 239 | 127 | |
BP315329.1 | - | 1 - 2 | 178 | 127 | |
CD704778.1 | - | 1 - 2 | 299 | 127 | |
DA070200.1 | - | 1 - 2 | 260 | 127 | |
DC403138.1 | - | 1 - 2 | 310 | 127 | |
U72514.1 | - | 1 - 2 | 153 | 127 |