The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052845.3 | NP_443077.1 | 4 - 5 | 422 | 57 |
NR_038118.1 | 4 - 5 | 422 | 57 | ||
XM_011538267.2 | XP_011536569.1 | 3 - 4 | 452 | 57 | |
XM_011538268.2 | XP_011536570.1 | 2 - 3 | 754 | 57 | |
XM_011538269.2 | XP_011536571.1 | 2 - 3 | 554 | 57 | |
GenBank mRNA | AK295211.1 | - | 5 - 6 | 530 | 57 |
AK312598.1 | - | 4 - 5 | 414 | 57 | |
BC005054.1 | - | 4 - 5 | 362 | 57 | |
BC011831.2 | - | 4 - 5 | 366 | 57 | |
DA718661.1 | - | 4 - 5 | 422 | 57 |