The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199629.1 | NP_001186558.1 | 7 - 8 | 954 | 90 |
NM_002475.4 | NP_002466.1 | 7 - 8 | 890 | 90 | |
GenBank mRNA | AI349553.1 | - | 1 - 2 | 90 | |
BC012425.1 | - | 6 - 7 | 90 | ||
BC014400.1 | - | 7 - 8 | 954 | 90 | |
M31211.1 | - | 6 - 7 | 658 | 90 | |
X16434.1 | - | 6 - 7 | 643 | 90 |