The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080476.4 | NP_536724.1 | 1 - 2 | 327 | 60 |
XM_017027664.1 | XP_016883153.1 | 1 - 2 | 145 | 60 | |
XM_017027665.1 | XP_016883154.1 | 1 - 2 | 145 | 60 | |
GenBank mRNA | AB086842.1 | - | 1 - 2 | 133 | 60 |
AK075507.1 | - | 1 - 2 | 152 | 60 | |
AK301900.1 | - | 1 - 2 | 127 | 60 | |
AK308627.1 | - | 1 - 2 | 134 | 60 | |
AY339061.1 | - | 1 - 2 | 127 | 60 | |
AY358816.1 | - | 1 - 2 | 157 | 60 | |
AY422169.1 | - | 1 - 2 | 327 | 60 | |
BC030512.1 | - | 1 - 2 | 138 | 60 |