Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYN3
Entrez Gene ID
8224
Gene Name
synapsin III
Gene Aliases
-
Chromosome Location
Chr.22: 32508022 - 33058391 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135774.1 NP_001129246.1 5 - 6 877 69
NM_003490.3 NP_003481.3 4 - 5 777 69
NM_133633.2 NP_598344.2 4 - 5 777 69
XM_011530405.2 XP_011528707.1 5 - 6 1413 69
XM_011530406.2 XP_011528708.1 5 - 6 879 69
XM_011530407.2 XP_011528709.1 5 - 6 1286 69
XM_011530408.1 XP_011528710.1 5 - 6 877 69
XM_011530410.2 XP_011528712.1 3 - 4 1081 69
XM_017028960.1 XP_016884449.1 6 - 7 1811 69
XM_017028961.1 XP_016884450.1 5 - 6 1203 69
XM_017028962.1 XP_016884451.1 5 - 6 1138 69
XM_017028963.1 XP_016884452.1 5 - 6 802 69
XM_017028964.1 XP_016884453.1 5 - 6 874 69
GenBank mRNA AB209684.1 - 5 - 6 829 69
AF046873.1 - 4 - 5 654 69
BC075065.2 - 4 - 5 644 69
BC075066.2 - 4 - 5 644 69
BC117460.1 - 4 - 5 642 69
BC143874.1 - 4 - 5 645 69
CR456589.1 - 4 - 5 709 69

View Full Product Details