Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYTL5
Entrez Gene ID
94122
Gene Name
synaptotagmin like 5
Gene Aliases
slp5
Chromosome Location
Chr.X: 37888915 - 38128820 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163334.1 NP_001156806.1 12 - 13 1857 77
NM_001163335.1 NP_001156807.1 13 - 14 1969 77
NM_138780.2 NP_620135.1 12 - 13 1878 77
XM_011544001.2 XP_011542303.1 15 - 16 2296 77
XM_011544002.2 XP_011542304.1 13 - 14 1930 77
XM_017029972.1 XP_016885461.1 16 - 17 2462 77
XM_017029973.1 XP_016885462.1 15 - 16 2529 77
XM_017029974.1 XP_016885463.1 14 - 15 2157 77
GenBank mRNA AB080222.1 - 11 - 12 1435 77
AK125106.1 - 12 - 13 1846 77
BC131585.1 - 12 - 13 1501 77
BX647688.1 - 13 - 14 1971 77

View Full Product Details