The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004691.4 | NP_004682.2 | 3 - 4 | 585 | 70 |
XM_011523444.1 | XP_011521746.1 | 4 - 5 | 572 | 70 | |
XM_011523445.1 | XP_011521747.1 | 4 - 5 | 705 | 70 | |
XM_011523446.1 | XP_011521748.1 | 3 - 4 | 477 | 70 | |
GenBank mRNA | AK300570.1 | - | 4 - 5 | 640 | 70 |
AK301630.1 | - | 4 - 5 | 574 | 70 | |
BC008861.2 | - | 3 - 4 | 563 | 70 | |
BI552108.1 | - | 3 - 4 | 599 | 70 | |
X71490.1 | - | 3 - 4 | 510 | 70 |