The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042581.1 | NP_001036046.1 | 4 - 5 | 745 | 81 |
NM_001042588.1 | NP_001036053.1 | 4 - 5 | 550 | 81 | |
NM_005701.3 | NP_005692.1 | 4 - 5 | 850 | 81 | |
GenBank mRNA | AF039029.1 | - | 4 - 5 | 495 | 81 |
AK054641.1 | - | 5 - 6 | 983 | 81 | |
AK289475.1 | - | 4 - 5 | 546 | 81 | |
BC004203.2 | - | 4 - 5 | 481 | 81 | |
CR456811.1 | - | 3 - 4 | 405 | 81 |