The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005259020.1 | XP_005259077.1 | 13 - 14 | 1584 | 81 |
XM_011527050.1 | XP_011525352.1 | 8 - 9 | 1201 | 81 | |
GenBank mRNA | AF282167.1 | - | 4 - 5 | 733 | 81 |
AK057052.1 | - | 4 - 5 | 881 | 81 | |
AK301615.1 | - | 3 - 4 | 544 | 81 | |
BC004907.2 | - | 10 - 11 | 1604 | 81 |