The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022070.4 | NP_071353.4 | 16 - 17 | 2569 | 112 |
XM_005257570.3 | XP_005257627.1 | 17 - 18 | 2757 | 112 | |
XM_011525110.2 | XP_011523412.1 | 15 - 16 | 2456 | 112 | |
XM_011525111.2 | XP_011523413.1 | 15 - 16 | 2418 | 112 | |
XM_017024936.1 | XP_016880425.1 | 16 - 17 | 2830 | 112 | |
XM_017024937.1 | XP_016880426.1 | 16 - 17 | 4506 | 112 | |
XM_017024938.1 | XP_016880427.1 | 16 - 17 | 2562 | 112 | |
XM_017024939.1 | XP_016880428.1 | 16 - 17 | 2833 | 112 | |
GenBank mRNA | AF349752.1 | - | 17 - 18 | 2514 | 112 |
AK025740.1 | - | 9 - 10 | 1636 | 112 | |
AK026070.1 | - | 9 - 10 | 1409 | 112 | |
AK127708.1 | - | 16 - 17 | 2563 | 112 | |
AK314655.1 | - | 16 - 17 | 2572 | 112 | |
BC119771.1 | - | 14 - 15 | 2223 | 112 | |
BC119813.1 | - | 16 - 17 | 2560 | 112 | |
BX640819.1 | - | 16 - 17 | 2560 | 112 | |
BX640831.1 | - | 16 - 17 | 2560 | 112 | |
CR627418.1 | - | 16 - 17 | 2559 | 112 |