The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286509.1 | NP_001273438.1 | 3 - 4 | 515 | 94 |
NM_001286510.1 | NP_001273439.1 | 3 - 4 | 514 | 94 | |
NM_001286511.1 | NP_001273440.1 | 3 - 4 | 469 | 94 | |
NM_001286512.1 | NP_001273441.1 | 3 - 4 | 518 | 94 | |
NM_001286517.1 | NP_001273446.1 | 2 - 3 | 325 | 94 | |
NM_178148.3 | NP_835361.1 | 3 - 4 | 519 | 94 | |
GenBank mRNA | AB097021.1 | - | 3 - 4 | 476 | 94 |
AB097039.1 | - | 3 - 4 | 476 | 94 | |
AB106538.1 | - | 3 - 4 | 367 | 94 | |
AK075456.1 | - | 3 - 4 | 448 | 94 | |
AK123749.1 | - | 3 - 4 | 416 | 94 | |
AK223028.1 | - | 2 - 3 | 169 | 94 | |
AY491520.1 | - | 3 - 4 | 485 | 94 | |
BC024288.1 | - | 3 - 4 | 483 | 94 | |
BI769501.1 | - | 2 - 3 | 255 | 94 | |
CN370928.1 | - | 3 - 4 | 415 | 94 | |
DA817588.1 | - | 3 - 4 | 469 | 94 | |
DB010939.1 | - | 3 - 4 | 450 | 94 |