Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP47
Entrez Gene ID
286464
Gene Name
cilia and flagella associated protein 47
Gene Aliases
CHDC2, CXorf22, CXorf30, CXorf59
Chromosome Location
Chr.X: 35919734 - 36385319 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304548.1 NP_001291477.1 7 - 8 1243 99
NM_152632.3 NP_689845.2 7 - 8 1243 99
XM_017029452.1 XP_016884941.1 7 - 8 1236 99
XM_017029453.1 XP_016884942.1 7 - 8 1236 99
GenBank mRNA AK126295.1 - 7 - 8 1209 99
BC027936.1 - 7 - 8 1196 99

View Full Product Details