The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320652.1 | NP_001307581.1 | 4 - 5 | 910 | 74 |
NM_001763.2 | NP_001754.2 | 4 - 5 | 1418 | 74 | |
GenBank mRNA | AK312945.1 | - | 4 - 5 | 1154 | 74 |
AW949640.1 | - | 1 - 2 | 96 | 74 | |
BC031645.1 | - | 6 - 7 | 996 | 74 | |
M27735.1 | - | 3 - 4 | 822 | 74 | |
M28825.1 | - | 4 - 5 | 1418 | 74 | |
X04450.1 | - | 3 - 4 | 591 | 74 |