The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032730.1 | NP_001027902.1 | 6 - 7 | 1006 | 111 |
NM_032236.5 | NP_115612.4 | 6 - 7 | 1006 | 111 | |
XM_005246009.2 | XP_005246066.1 | 6 - 7 | 1010 | 111 | |
XM_006710951.2 | XP_006711014.2 | 6 - 7 | 1010 | 111 | |
XM_006710955.3 | XP_006711018.1 | 6 - 7 | 986 | 111 | |
XM_011542262.2 | XP_011540564.1 | 6 - 7 | 903 | 111 | |
XM_011542263.2 | XP_011540565.1 | 6 - 7 | 903 | 111 | |
XM_011542264.1 | XP_011540566.1 | 5 - 6 | 600 | 111 | |
XM_011542265.2 | XP_011540567.1 | 7 - 8 | 1566 | 111 | |
XM_011542267.2 | XP_011540569.1 | 6 - 7 | 903 | 111 | |
XM_017002495.1 | XP_016857984.1 | 6 - 7 | 1010 | 111 | |
XM_017002496.1 | XP_016857985.1 | 6 - 7 | 1010 | 111 | |
GenBank mRNA | AF502942.1 | - | 6 - 7 | 908 | 111 |
AK074247.1 | - | 6 - 7 | 903 | 111 | |
AK226150.1 | - | 6 - 7 | 1002 | 111 | |
AK315005.1 | - | 6 - 7 | 786 | 111 | |
BC067261.1 | - | 6 - 7 | 974 | 111 | |
BC103724.1 | - | 6 - 7 | 948 | 111 | |
BC104896.1 | - | 6 - 7 | 802 | 111 | |
BC143343.1 | - | 6 - 7 | 802 | 111 | |
BC143349.1 | - | 6 - 7 | 802 | 111 | |
BC143354.1 | - | 6 - 7 | 802 | 111 |