Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAUS5
Entrez Gene ID
23354
Gene Name
HAUS augmin like complex subunit 5
Gene Aliases
KIAA0841, dgt5
Chromosome Location
Chr.19: 35612744 - 35625349 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015302.1 NP_056117.1 3 - 4 238 99
GenBank mRNA AB020648.1 - 3 - 4 211 99
AK307703.1 - 3 - 4 228 99
BC013947.2 - 3 - 4 210 99
BC064390.1 - 3 - 4 238 99
BC157882.1 - 3 - 4 229 99
BC171905.1 - 3 - 4 230 99

View Full Product Details