Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAUS5
Entrez Gene ID
23354
Gene Name
HAUS augmin like complex subunit 5
Gene Aliases
KIAA0841, dgt5
Chromosome Location
Chr.19: 35612744 - 35625349 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015302.1 NP_056117.1 10 - 11 864 100
XM_011526684.2 XP_011524986.1 9 - 10 751 100
GenBank mRNA AB020648.1 - 10 - 11 837 100
BC013947.2 - 10 - 11 836 100
BC157882.1 - 10 - 11 855 100
BC171905.1 - 10 - 11 856 100

View Full Product Details