The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032437.3 | NP_115813.2 | 4 - 5 | 819 | 79 |
XM_005271274.3 | XP_005271331.1 | 4 - 5 | 619 | 79 | |
XM_006710976.3 | XP_006711039.1 | 4 - 5 | 713 | 79 | |
XM_006710977.1 | XP_006711040.1 | 4 - 5 | 741 | 79 | |
XM_011542301.2 | XP_011540603.1 | 4 - 5 | 734 | 79 | |
XM_017002548.1 | XP_016858037.1 | 4 - 5 | 574 | 79 | |
GenBank mRNA | AB058702.1 | - | 4 - 5 | 582 | 79 |
AK092219.1 | - | 4 - 5 | 730 | 79 | |
AK292220.1 | - | 4 - 5 | 778 | 79 | |
AK315850.1 | - | 4 - 5 | 595 | 79 | |
BC015814.1 | - | 4 - 5 | 523 | 79 |