Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC28
Entrez Gene ID
23331
Gene Name
tetratricopeptide repeat domain 28
Gene Aliases
TPRBK
Chromosome Location
Chr.22: 27978014 - 28679865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145418.1 NP_001138890.1 9 - 10 3558 83
XM_005261405.2 XP_005261462.1 9 - 10 3558 83
XM_006724171.3 XP_006724234.1 8 - 9 12336 83
XM_011530018.2 XP_011528320.1 9 - 10 3737 83
XM_011530019.2 XP_011528321.1 9 - 10 3558 83
XM_011530020.1 XP_011528322.1 9 - 10 3558 83
XM_011530021.2 XP_011528323.1 9 - 10 3558 83
XM_011530022.1 XP_011528324.1 9 - 10 3558 83
XM_017028673.1 XP_016884162.1 9 - 10 3531 83
GenBank mRNA AB028966.3 - 4 - 5 2351 83
AB665284.1 - 9 - 10 3506 83
BC016465.1 - 1 - 2 102 83

View Full Product Details