Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRMT1L
Entrez Gene ID
81627
Gene Name
tRNA methyltransferase 1 like
Gene Aliases
C1orf25, MST070, MSTP070, TRM1L, bG120K12.3
Chromosome Location
Chr.1: 185118085 - 185157098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202423.1 NP_001189352.1 9 - 10 1201 92
NM_030934.4 NP_112196.3 9 - 10 1593 92
XM_011510028.2 XP_011508330.1 9 - 10 3030 92
XM_017002419.1 XP_016857908.1 9 - 10 1565 92
GenBank mRNA AF163261.1 - 1 - 2 56 92
AF288399.1 - 9 - 10 1515 92
AK074936.1 - 9 - 10 1542 92
AK074993.1 - 9 - 10 1565 92
AK098359.1 - 9 - 10 1560 92
AK131460.1 - 8 - 9 2235 92
AK226103.1 - 9 - 10 1308 92
AK302166.1 - 10 - 11 1125 92
AK312801.1 - 9 - 10 1562 92
BC035430.1 - 9 - 10 1545 92
BC039738.1 - 9 - 10 1556 92
BC045535.1 - 9 - 10 1542 92
BC053961.1 - 9 - 10 1542 92

View Full Product Details