Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPON1
Entrez Gene ID
10418
Gene Name
spondin 1
Gene Aliases
VSGP/F-spondin, f-spondin
Chromosome Location
Chr.11: 13962637 - 14268133 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006108.3 NP_006099.2 8 - 9 1367 145
GenBank mRNA AB018305.2 - 7 - 8 852 145
AB051390.1 - 8 - 9 1123 145
AK074803.1 - 8 - 9 1313 145
AK291780.1 - 8 - 9 1315 145
BC136513.1 - 8 - 9 1309 145
BC136563.1 - 8 - 9 1309 145

View Full Product Details