Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TXNDC16
Entrez Gene ID
57544
Gene Name
thioredoxin domain containing 16
Gene Aliases
ERp90, KIAA1344
Chromosome Location
Chr.14: 52430590 - 52552583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160047.1 NP_001153519.1 10 - 11 1353 102
NM_020784.2 NP_065835.2 10 - 11 1368 102
XM_017021505.1 XP_016876994.1 10 - 11 1348 102
GenBank mRNA AB037765.2 - 10 - 11 1280 102
AK097748.1 - 10 - 11 1155 102
BC137081.1 - 10 - 11 1248 102
BC137082.1 - 10 - 11 1248 102
BC142650.1 - 10 - 11 1291 102
BC144469.1 - 10 - 11 1233 102
BC150205.1 - 10 - 11 1289 102
BC152428.1 - 10 - 11 1280 102

View Full Product Details