Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC16
Entrez Gene ID
57544
Gene Name
thioredoxin domain containing 16
Gene Aliases
ERp90, KIAA1344
Chromosome Location
Chr.14: 52430590 - 52552583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160047.1 NP_001153519.1 16 - 17 2053 150
NM_020784.2 NP_065835.2 16 - 17 2068 150
GenBank mRNA AB037765.2 - 16 - 17 1980 150
AK022563.1 - 2 - 3 254 150
AK097748.1 - 17 - 18 1957 150
BC137081.1 - 16 - 17 1948 150
BC137082.1 - 16 - 17 1948 150
BC142650.1 - 16 - 17 1991 150
BC144469.1 - 16 - 17 1933 150
BC150205.1 - 16 - 17 1989 150
BC152428.1 - 16 - 17 1980 150

View Full Product Details