Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC16
Entrez Gene ID
57544
Gene Name
thioredoxin domain containing 16
Gene Aliases
ERp90, KIAA1344
Chromosome Location
Chr.14: 52430590 - 52552583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160047.1 NP_001153519.1 19 - 20 2430 111
NM_020784.2 NP_065835.2 19 - 20 2445 111
GenBank mRNA AB037765.2 - 19 - 20 2357 111
AK022563.1 - 5 - 6 631 111
AK097748.1 - 20 - 21 2334 111
BC137081.1 - 19 - 20 2325 111
BC137082.1 - 19 - 20 2325 111
BC142650.1 - 19 - 20 2368 111
BC144469.1 - 19 - 20 2310 111
BC150205.1 - 19 - 20 2366 111
BC152428.1 - 19 - 20 2357 111

View Full Product Details