Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIP2A
Entrez Gene ID
23181
Gene Name
disco interacting protein 2 homolog A
Gene Aliases
C21orf106, DIP2
Chromosome Location
Chr.21: 46458942 - 46570900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146115.1 NP_001139587.1 12 - 13 1697 58
NM_001146116.1 NP_001139588.1 13 - 14 1814 58
NM_015151.3 NP_055966.2 13 - 14 1826 58
NM_206889.2 NP_996772.1 13 - 14 1826 58
NM_206890.2 NP_996773.1 13 - 14 1826 58
NM_206891.2 NP_996774.1 13 - 14 1826 58
XM_011529492.2 XP_011527794.1 14 - 15 1941 58
XM_011529501.1 XP_011527803.1 14 - 15 1941 58
XM_011529502.1 XP_011527804.1 14 - 15 1941 58
XM_017028291.1 XP_016883780.1 14 - 15 1941 58
XM_017028292.1 XP_016883781.1 14 - 15 1941 58
XM_017028293.1 XP_016883782.1 14 - 15 1938 58
XM_017028294.1 XP_016883783.1 13 - 14 1820 58
XM_017028295.1 XP_016883784.1 13 - 14 1817 58
XM_017028296.1 XP_016883785.1 13 - 14 1817 58
XM_017028297.1 XP_016883786.1 13 - 14 1809 58
XM_017028298.1 XP_016883787.1 12 - 13 1688 58
XM_017028299.1 XP_016883788.1 13 - 14 1686 58
XM_017028300.1 XP_016883789.1 14 - 15 1941 58
XM_017028301.1 XP_016883790.1 14 - 15 1941 58
XM_017028302.1 XP_016883791.1 12 - 13 1581 58
XM_017028303.1 XP_016883792.1 14 - 15 1941 58
XM_017028304.1 XP_016883793.1 14 - 15 1941 58
XM_017028305.1 XP_016883794.1 14 - 15 1941 58
GenBank mRNA AB273729.1 - 13 - 14 1664 58
AF432263.1 - 8 - 9 1036 58
AF490768.1 - 13 - 14 1719 58
AK056738.1 - 13 - 14 1718 58
AK098236.1 - 8 - 9 969 58
AK303351.1 - 11 - 12 1641 58
BC033718.1 - 12 - 13 1680 58
BC038443.1 - 13 - 14 1825 58

View Full Product Details