The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017539.2 | NP_060009.1 | 12 - 13 | 1815 | 148 |
XM_006721062.1 | XP_006721125.1 | 12 - 13 | 1911 | 148 | |
XM_011545882.2 | XP_011544184.1 | 13 - 14 | 2351 | 148 | |
XM_011545883.1 | XP_011544185.1 | 13 - 14 | 2119 | 148 | |
XM_011545885.2 | XP_011544187.1 | 13 - 14 | 2351 | 148 | |
XM_011545886.2 | XP_011544188.1 | 5 - 6 | 726 | 148 | |
XM_017023426.1 | XP_016878915.1 | 13 - 14 | 2351 | 148 | |
XM_017023428.1 | XP_016878917.1 | 13 - 14 | 2351 | 148 | |
XM_017023429.1 | XP_016878918.1 | 13 - 14 | 2352 | 148 | |
XM_017023431.1 | XP_016878920.1 | 13 - 14 | 2352 | 148 | |
XM_017023432.1 | XP_016878921.1 | 12 - 13 | 1911 | 148 | |
GenBank mRNA | AF494040.1 | - | 12 - 13 | 1815 | 148 |
AK056509.2 | - | 12 - 13 | 1911 | 148 | |
AK303958.1 | - | 12 - 13 | 1716 | 148 |